Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Brand: RETSCH 60.167.045000
6806.75 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.60000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel | |
65mm |
For separation, fractioning, particle size determination | |
400 mm | |
45mm | |
ASTM E 11 | |
Shaker AS 450 |