Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve with Square Holes, ISO Certified
Brand: RETSCH 60.154.056000
6236.75 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 2.00000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel |
For separation, fractioning, particle size determination | |
400 mm | |
56mm | |
ISO 3310/2 | |
65mm |