Learn More
Jenway™ 4520 Conductivity Meter without Probe
Offers accuracy with 1, 2 or 3 point conductivity calibration across the wide measurement range of 0 to 19.99S. Jenway™ 4520 Conductivity Meter without Probe ensures optimum accuracy with a dedicated “pure water” mode.
Brand: Jenway™ 452000-BI
Additional Details : Weight : 0.85000kg
Description
- Conductivity, resistivity, TDS and salinity modes
- Special pure water mode for ultra low conductivity measurements
- Storage of 500 results with built in data logger
- Infra-red data link (IrDA) for connection to printer and RS232 link to connect to printer or PC via DataWay software
- 3 year warranty
Specifications
±0.5% ±2 digits | |
±0.5% ±2 digits | |
0.010 to 19.999 | |
210 x 250 x 55 mm | |
UK | |
0.01 μS to 0.01 S | |
0.1 g/L | |
0.1°C | |
9V | |
±0.5% ±2 digits | |
0 to 20 Mohm | |
6 auto selected ranges from 0 to 1999 g/L |
0 to 35 ±1 g/L, 35 to 99.9 ±3 g/L | |
±0.5°C | |
Conductivity Meter without Probe | |
9V AC ±10%, 50/60Hz | |
Not included | |
0.01 Mohm | |
0.01 mg/L to 1 g/L | |
18, 20, 25°C | |
850 g | |
6 auto selected from 0 to 19.99 S* | |
0 to 99.9 g/L | |
-10°C to +105°C |