Läs mer
4a-Methyl-cholesterol, TRC
Produktkod. 30137307
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137307 100 mg
30136151 50 mg
30006331 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137307 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM295715100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15073-00-4
Molekylformel C28H48O
Molekylvikt (g/mol) 400.68
Synonym 4a-Methyl-cholest-5-en-3b-ol, (3S,4S,8S,9S,10R,13R,14S,17R)-4,10,13-Trimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
IUPAC-namn (3S,4S,8S,9S,10R,13R,14S,17R)-4,10,13-trimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER O[C@H]([C@H]1C)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](CCCC(C)C)C
Specifikationer
Kemiskt namn eller material 4alpha-Methyl-cholest-5-en-3beta-ol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C28H48O/c1-18(2)8-7-9-19(3)22-12-13-24-21-10-11-23-20(4)26(29)15-17-28(23,6)25(21)14-16-27(22,24)5/h11,18-22,24-26,29H,7-10,12-17H2,1-6H3/t19-,20+,21+,22-,24+,25+,26+,27-,28+/m1/s1
Formel vikt 400.3705
Procent renhet >95%