Läs mer
4Alpha-Hydroxy Cholesterol, TRC
Produktkod. 30010760
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010760 10 mg
30009114 5 mg
30005411 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH91797510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34310-86-6
Molekylformel C27 H46 O2
Molekylvikt (g/mol) 402.65
Synonym Cholest-5-ene-3,4-diol, (3β,4α)-, Cholest-5-ene-3β,4α-diol (8CI), (3β,4α)-Cholest-5-ene-3,4-diol, 4α-Hydroxycholesterol, 4α-Hydroxy Cholesterol, (3β,4α)-Cholest-5-ene-3,4-diol, 3β,4α-Dihydroxycholest-5-ene, trans-3,4-Dihydroxy-δ5-cholestene, trans-δ5-Cholestene-3,4-diol
IUPAC-namn (3S,4S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-3,4-diol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4[C@H](O)[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 4alpha-Hydroxycholesterol
Kvantitet 10 mg
InChI formel InChI=1S/C27H46O2/c1-17(2)7-6-8-18(3)20-11-12-21-19-9-10-23-25(29)24(28)14-16-27(23,5)22(19)13-15-26(20,21)4/h10,17-22,24-25,28-29H,6-9,11-16H2,1-5H3/t18-,19+,20-,21+,22+,24+,25+,26-,27-/m1/s1
Formel vikt 402.3498