Läs mer
(4E, 14Z)-Sphingadienine-C18-1-phosphate, TRC
Produktkod. 30006561
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006561 1 mg
30012330 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006561 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6806351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1622300-16-6
Molekylformel C18H36NO5P
Molekylvikt (g/mol) 377.46
Synonym (2S,3R,4E,14Z)-2-Amino-3-hydroxyoctadeca-4,14-dien-1-yl Dihydrogen Phosphate, (E,Z)-D-erythro-2-Amino-4,14-octadecadiene-1,3-diol Dihydrogen Phosphate, [R-[R*,S*-(E,Z)]]-2-Amino-4,14-octadecadiene-1,3-diol Dihydrogen Phosphate, Sphingadienine Dihydrogen Phosphate, cis-4,14-Sphingadienine Dihydrogen Phosphate
LEDER OCCOCCOc1ccc(cc1)C(CC)(C)CCC(C)C
Specifikationer
Kemiskt namn eller material (4E, 14Z)-Sphingadienine-C18-1-phosphate
Kvantitet 1 mg
InChI formel InChI=1S/C19H32O3/c1-5-19(4,11-10-16(2)3)17-6-8-18(9-7-17)22-15-14-21-13-12-20/h6-9,16,20H,5,10-15H2,1-4H3