Läs mer
(4E, 8E)-Sphingadienine-C18-1-phosphate, TRC
Produktkod. 30409325
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30409325 1 mg
1 options
Produktkod. 30409325 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6806801MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4E','8E)-Sphingadienine-C18-1-phosphate
CAS 1419705-13-7
Molekylformel C23 H32 O6
Kvantitet 1 mg
Synonym (2S,3R,4E)-2-Amino-4,8-octadecadiene-1,3-diol 1-(Dihydrogen phosphate), (2S,3R,4E,8E)-2-Amino-3-hydroxyoctadeca-4,8-dien-1-yl Dihydrogen Phosphate
InChI formel InChI=1S/C23H32O6/c1-20-7-5-15(24)9-14(20)3-4-16-17-6-8-22(21(17,2)10-18(25)19(16)20)23(29-13-27-22)11-26-12-28-23/h9,16-19,25H,3-8,10-13H2,1-2H3/t16-,17-,18-,19+,20-,21-,22+,23?/m0/s1
LEDER C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@@]5(OCOC56COCO6)[C@@]4(C)C[C@H](O)[C@H]23
Molekylvikt (g/mol) 404.497
Formel vikt 404.22