Läs mer
(4R)-1-Methyl-4-propyl-L-proline Methyl Ester, TRC
Produktkod. 30468538
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30468538 5 mg
1 options
Produktkod. 30468538 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3202905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4R)-1-Methyl-4-propyl-L-proline Methyl Ester
CAS 13380-39-7
Molekylformel C10H19NO2
Kvantitet 5 mg
Synonym 4-Propyl-hygric Acid Methyl Ester, (4R)-4-Propyl-hygric Acid Methyl Ester, (2S,4R)-4-Propyl-1-methylpyrrolidine-2-carboxylic Acid Methyl Ester, trans-4-Propyl-1methyl-L-proline Methyl Ester
InChI formel InChI=1S/C10H19NO2/c1-4-5-8-6-9(10(12)13-3)11(2)7-8/h8-9H,4-7H2,1-3H3/t8-,9+/m1/s1
LEDER CCC[C@@H]1C[C@H](N(C)C1)C(=O)OC
IUPAC-namn methyl (2S,4R)-1-methyl-4-propylpyrrolidine-2-carboxylate
Molekylvikt (g/mol) 185.26
Formel vikt 185.1416