Läs mer
(4R)-(3’,4’-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one, TRC
Produktkod. 30012807
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012807 100 mg
30009153 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD435708100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 155748-61-1
Molekylformel C16 H12 Cl2 O
Molekylvikt (g/mol) 291.17
Synonym (4R)-4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, 1(2H)-Naphthalenone, 4-(3,4-dichlorophenyl)-3,4-dihydro-, (R)-, (4R)-4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, (4R)-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, (R)-4-(3,4-Dichlorophenyl)-1-tetralone, (R)-4-(3,4-Dichlorophenyl)-3,4-dihydro-1-naphthalenone, (4R)-4-(3,4-dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one, Sertraline Imp. F (EP)
IUPAC-namn (4R)-4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
LEDER Clc1ccc(cc1Cl)[C@H]2CCC(=O)c3ccccc23
Specifikationer
Kemiskt namn eller material (4R)-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Kvantitet 100 mg
InChI formel InChI=1S/C16H12Cl2O/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13/h1-5,7,9,11H,6,8H2/t11-/m1/s1
Formel vikt 290.0265