Läs mer
(4R)-4-(1-Methylethyl)-2-cyclohexen-1-one (~80% ee), TRC
Produktkod. 30469078
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469078 100 mg
30473469 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469078 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM223865100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4R)-4-(1-Methylethyl)-2-cyclohexen-1-one
CAS 2158-59-0
Molekylformel C9H14O
Kvantitet 100 mg
Synonym (R)-4- Isopropylcyclohex-2-enone, (R)-4-(1-methylethyl)-2-cyclohexen-1-one, (R)-(-)- (8CI)-4-isopropyl-2-cyclohexen-1-one, (4R)-4-(1-Methylethyl)-2-cyclohexen-1-one, (R)-(-)-Cryptone, l-Cryptone
InChI formel InChI=1S/C9H14O/c1-7(2)8-3-5-9(10)6-4-8/h3,5,7-8H,4,6H2,1-2H3/t8-/m1/s1
LEDER CC(C)[C@H]1CCC(=O)C=C1
IUPAC-namn (4R)-4-propan-2-ylcyclohex-2-en-1-one
Molekylvikt (g/mol) 138.21
Formel vikt 138.1045