Läs mer
(4R-5R)-4,5-Dihydro-5-[4-(methylsulfonyl)phenyl]-2-phenyl-4-oxazolecarboxylic Acid Ethyl Ester, TRC
Produktkod. 30387728
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30387728 250 mg
1 options
Produktkod. 30387728 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD449225250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4R-5R)-4,5-Dihydro-5-[4-(methylsulfonyl)phenyl]-2-phenyl-4-oxazolecarboxylic Acid Ethyl Ester
CAS 139059-00-0
Molekylformel C19H19NO5S
Kvantitet 250 mg
Synonym (4R-cis)-4,5-Dihydro-5-[4-(methylsulfonyl)phenyl]-2-phenyl-4-oxazolecarboxylic Acid Ethyl Ester, Ethyl (4R,5R)-D-erythro-4,5-Dihydro-5-(4-methylsulfonylphenyl)-2-phenyl-4-oxazolecarboxylate
InChI formel InChI=1S/C19H19NO5S/c1-3-24-19(21)16-17(13-9-11-15(12-10-13)26(2,22)23)25-18(20-16)14-7-5-4-6-8-14/h4-12,16-17H,3H2,1-2H3/t16-,17-/m1/s1
LEDER CCOC(=O)[C@@H]1N=C(O[C@@H]1c2ccc(cc2)S(=O)(=O)C)c3ccccc3
IUPAC-namn ethyl (4R,5R)-5-(4-methylsulfonylphenyl)-2-phenyl-4,5-dihydro-1,3-oxazole-4-carboxylate
Molekylvikt (g/mol) 373.42
Formel vikt 373.0984