missing translation for 'onlineSavingsMsg'
Learn More

(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone, 99%, Thermo Scientific Chemicals

Catalog Number p-3779183
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
77943-39-6
C10H11NO2
177.2
MFCD00010845
PPIBJOQGAJBQDF-VXNVDRBHSA-N
4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, meph-2-oxazolidinone, pubchem6871, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone
2733812
(4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one
CC1C(OC(=O)N1)C2=CC=CC=C2
This item is not returnable. View return policy
77943-39-6
C10H11NO2
177.2
MFCD00010845
PPIBJOQGAJBQDF-VXNVDRBHSA-N
4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, meph-2-oxazolidinone, pubchem6871, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone
2733812
(4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one
CC1C(OC(=O)N1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 121°C
CAS Min % 98.5
CAS Max % 100.0
Color Brown-Yellow to Colorless
Assay Percent Range 98.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation + 162
Formula Weight 177.2
Percent Purity 99%
Specific Rotation Condition + 162.00 (20.00°C c=2,CHCL3)
Quantity 1 g
Physical Form Liquid
Chemical Name or Material (4R, 5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'