Läs mer
(4R,7S)Tedalinab, TRC
Produktkod. 30456779
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456779 1 mg
30450026 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456779 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3579101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 916591-02-1
Molekylformel C19H21F2N3O
Molekylvikt (g/mol) 345.39
Synonym (4R,7S)-1-(2,4-Difluorophenyl)-N-(1,1-dimethylethyl)-4,5,6,7-tetrahydro-4,7-methano-1H-indazole-3-carboxamide
LEDER CC(C)(C)NC(=O)c1nn(c2[C@H]3CC[C@H](C3)c12)c4ccc(F)cc4F
Specifikationer
Kemiskt namn eller material (4R,7S)Tedalinab
Kvantitet 1 mg
InChI formel InChI=1S/C19H21F2N3O/c1-19(2,3)22-18(25)16-15-10-4-5-11(8-10)17(15)24(23-16)14-7-6-12(20)9-13(14)21/h6-7,9-11H,4-5,8H2,1-3H3,(H,22,25)/t10-,11+/m1/s1
Formel vikt 345.1653