Läs mer
(4S)-2,2,4-Trimethylpyrrolidine Hydrochloride, TRC
Produktkod. 30454584
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454584 100 mg
30449476 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT787390100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1897428-40-8
Molekylformel C7H15N . HCl
Molekylvikt (g/mol) 149.66
Synonym (S)-2,2,4-Trimethylpyrrolidine Hydrochloride
IUPAC-namn (4S)-2,2,4-trimethylpyrrolidine;hydrochloride
LEDER Cl.C[C@@H]1CNC(C)(C)C1
Specifikationer
Kemiskt namn eller material (4S)-2,2,4-Trimethylpyrrolidine Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C7H15N.ClH/c1-6-4-7(2,3)8-5-6;/h6,8H,4-5H2,1-3H3;1H/t6-;/m0./s1
Formel vikt 149.0971