Läs mer
(4S)-2,5-Dioxo-4-oxazolidinepropanoic Acid Phenylmethyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (4S)-2,5-Dioxo-4-oxazolidinepropanoic Acid Phenylmethyl Ester |
| CAS | 3190-71-4 |
| Molekylformel | C13 H13 N O5 |
| Kvantitet | 1 g |
| Synonym | (+)-(S)-Glutamic Acid γ-Benzyl Ester N-carboxyanhydride, 5-Benzyl L-Glutamate N-Carboxyanhydride, L-Glutamic Acid-γ-benzyll ester N-Carboxyanhydride, L-γ-Benzylglutamic Acid Carboxyanhydride, N-Carboxy-γ-benzyl-L-glutamate Anhydride, γ-Benzyl L-N-Carboxyglutamic Anhydride, γ-Benzyl L-Glutamate N-Carboxyanhydride, γ-Benzyl L-Glutamate-N-carboxylic anhydride, γ-Benzyl N-carboxy-L-glutamic Anhydride, γ-Benzyl Glutamate N-Carboxyanhydride, γ-Benzyl Glutamate NCA, γ-Benzyl-L-glutamic Acid Carboxyanhydride |
| InChI formel | InChI=1S/C13H13NO5/c15-11(18-8-9-4-2-1-3-5-9)7-6-10-12(16)19-13(17)14-10/h1-5,10H,6-8H2,(H,14,17)/t10-/m0/s1 |
| LEDER | O=C(CC[C@@H]1NC(=O)OC1=O)OCc2ccccc2 |
| IUPAC-namn | benzyl 3-[(4S)-2,5-dioxo-1,3-oxazolidin-4-yl]propanoate |
| Molekylvikt (g/mol) | 263.25 |
| Formel vikt | 263.0794 |