Läs mer
(4S)-4-Cyclohexyl-L-proline Hydrochloride, TRC
Produktkod. 30362806
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30362806 100 mg
1 options
Produktkod. 30362806 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC988110100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fosinopril Impurity C
CAS 90657-55-9
Molekylformel C11 H19 N O2 . Cl H
Kvantitet 100 mg
Synonym L-Proline, 4-cyclohexyl-, hydrochloride (1:1), (4S)-, L-Proline, 4-cyclohexyl-, hydrochloride, (4S)- (9CI), L-Proline, 4-cyclohexyl-, hydrochloride, trans-, trans-4-Cyclohexyl-L-proline hydrochloride
InChI formel InChI=1S/C11H19NO2.ClH/c13-11(14)10-6-9(7-12-10)8-4-2-1-3-5-8;/h8-10,12H,1-7H2,(H,13,14);1H/t9-,10+;/m1./s1
LEDER Cl.OC(=O)[C@@H]1C[C@H](CN1)C2CCCCC2
IUPAC-namn (2S,4S)-4-cyclohexylpyrrolidine-2-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 233.74
Formel vikt 233.1183