Läs mer
(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic Acid, Mikromol™
Produktkod. 30050193
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050193 100 mg
30049291 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050193 Leverantör Mikromol Leverantörsnummer MM3396.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic Acid
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazoli
CAS 143527-70-2
Analyt- eller komponentnamn (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazoli
Frakt skick Room Temperature
Molekylformel C17 H23 N O5
Certifikat och efterlevnad ISO 17025
Synonym 3,5-Oxazolidinedicarboxylic acid, 2,2-dimethyl-4-phenyl-, 3-(1,1-dimethylethyl) ester, (4S,5R)-, 3,5-Oxazolidinedicarboxylic acid, 2,2-dimethyl-4-phenyl-, 3-(1,1-dimethylethyl) ester, (4S-trans)-, (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-5-oxazolidinecarboxylic acid, (4S,5R)-3-tert-Butoxycarbonyl-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic acid, (4S,5R)-5-Carboxy-2,2-dimethyl-4-phenyl-3-(tert-butoxycarbonyl)oxazolidine, (4S,5R)-N-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-5-oxazolidinecarboxylic acid
InChI formel InChI=1S/C17H23NO5/c1-16(2,3)23-15(21)18-12(11-9-7-6-8-10-11)13(14(19)20)22-17(18,4)5/h6-10,12-13H,1-5H3,(H,19,20)/t12-,13+/m0/s1
LEDER CC(C)(C)OC(=O)N1[C@H]([C@@H](OC1(C)C)C(=O)O)c2ccccc2
IUPAC-namn (4S,5R)-2,2-dimethyl-3-[(2-methylpropan-2-yl)oxycarbonyl]-4-phenyl-1,3-oxazolidine-5-carboxylic acid
Molekylvikt (g/mol) 321.37
Kvantitet 100 mg
Formel vikt 321.1576 g/mol