Läs mer
(4S,5S)-2,2,5-Trimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, TRC
Produktkod. 30450140
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30450140 100 mg
1 options
Produktkod. 30450140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT796310100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4S,5S)-2,2,5-Trimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester
CAS 117181-61-0
Molekylformel C8H14O4
Kvantitet 100 mg
Synonym (4S-cis)-2,2,5-Trimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, (4S,5S)-Methyl 2,2,5-Trimethyl-1,3-dioxolane-4-carboxylate
InChI formel InChI=1S/C8H14O4/c1-5-6(7(9)10-4)12-8(2,3)11-5/h5-6H,1-4H3/t5-,6-/m0/s1
LEDER COC(=O)[C@H]1OC(C)(C)O[C@H]1C
IUPAC-namn methyl (4S,5S)-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate
Molekylvikt (g/mol) 174.19
Formel vikt 174.0892