Läs mer
(4S,cis)-1,1-Dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, TRC
Produktkod. 30368451
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30368451 1 mg -
30350321 2.5 mg 2.5mg
30357634 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368451 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4972491MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 947586-93-8
Molekylformel C14H27NO4
Molekylvikt (g/mol) 273.37
Synonym (4S,6S)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, [(4S,6S)-6-(2-Aminoethyl)-2,2-dimethyl[1,3]dioxan-4-yl]acetic Acid tert-Butyl Ester
IUPAC-namn tert-butyl 2-[(4S,6S)-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
LEDER CC1(O[C@H](C[C@H](O1)CC(=O)OC(C)(C)C)CCN)C
Specifikationer
Kemiskt namn eller material (4S,cis)-1,1-Dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
Kvantitet 1 mg
InChI formel InChI=1S/C14H27NO4/c1-13(2,3)19-12(16)9-11-8-10(6-7-15)17-14(4,5)18-11/h10-11H,6-9,15H2,1-5H3/t10-,11-/m0/s1