missing translation for 'onlineSavingsMsg'
Learn More

(4S)-(-)-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals

Catalog Number p-7021622
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
17016-83-0
C6H11NO2
129.159
MFCD00010847
YBUPWRYTXGAWJX-RXMQYKEDSA-N
s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
7157133
(4S)-4-propan-2-yl-1,3-oxazolidin-2-one
CC(C)C1COC(=O)N1
This item is not returnable. View return policy
17016-83-0
C6H11NO2
129.159
MFCD00010847
YBUPWRYTXGAWJX-RXMQYKEDSA-N
s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
7157133
(4S)-4-propan-2-yl-1,3-oxazolidin-2-one
CC(C)C1COC(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C to 74°C
Beilstein 3588198
Formula Weight 129.16
Percent Purity 98%
Quantity 1 g
Optical Rotation −17° (c=6 in Ethanol)
Chemical Name or Material (4S)-(-)-Isopropyl-2-oxazolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'