Läs mer
5-([1,1’-Biphenyl]-4-ylphenylmethyl)-1H-imidazole, TRC
Produktkod. 30133942
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133942 5 mg
30007587 100 mg
30005501 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133942 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3979255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91679-37-7
Molekylformel C22H18N2
Molekylvikt (g/mol) 310.39
Synonym 4-([1,1'-Biphenyl]-4-ylphenylmethyl)1H-imidazole, Bifonazole EP Impurity B;
IUPAC-namn 5-[phenyl-(4-phenylphenyl)methyl]-1H-imidazole
LEDER C(C1=CC=CC=C1)(C2=CNC=N2)C3=CC=C(C4=CC=CC=C4)C=C3
Specifikationer
Kemiskt namn eller material 5-([1,1'-Biphenyl]-4-ylphenylmethyl)-1H-imidazole
Kvantitet 5 mg
InChI formel InChI=1S/C22H18N2/c1-3-7-17(8-4-1)18-11-13-20(14-12-18)22(21-15-23-16-24-21)19-9-5-2-6-10-19/h1-16,22H,(H,23,24)
Formel vikt 310.147