Läs mer
5-(1,1-Dimethylheptyl)resorcinol, TRC
Produktkod. 30379335
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379335 1 g
30379304 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379335 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4731201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56469-10-4
Molekylformel C15H24O2
Molekylvikt (g/mol) 236.35
Synonym 5-(1,1-Dimethylheptyl)-1,3-benzenediol
LEDER CCCCCCC(C)(C)c1cc(O)cc(O)c1
Specifikationer
Kemiskt namn eller material 5-(1,1-Dimethylheptyl)resorcinol
Kvantitet 1 g
InChI formel InChI=1S/C15H24O2/c1-4-5-6-7-8-15(2,3)12-9-13(16)11-14(17)10-12/h9-11,16-17H,4-8H2,1-3H3
Formel vikt 236.1776