Läs mer
5-(1-Piperazinyl)-2-benzofurancarboxylic Acid Ethyl Ester, TRC
Produktkod. 30419184
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30419184 1 g
1 options
Produktkod. 30419184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4806351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-(1-Piperazinyl)-2-benzofurancarboxylic Acid Ethyl Ester
CAS 163521-20-8
Molekylformel C15H18N2O3
Kvantitet 1 g
Synonym Ethyl 5-(Piperazin-1-yl)benzofuran-2-carboxylate, Ethyl 5-(Piperazine-1-yl)-1-benzofuran-2-carboxylate
InChI formel InChI=1S/C15H18N2O3/c1-2-19-15(18)14-10-11-9-12(3-4-13(11)20-14)17-7-5-16-6-8-17/h3-4,9-10,16H,2,5-8H2,1H3
LEDER CCOC(=O)c1oc2ccc(cc2c1)N3CCNCC3
Molekylvikt (g/mol) 274.32