Läs mer
Δ-5(10)-Norethindrone Acetate, TRC
Produktkod. 30412464
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30412464 10 mg
1 options
Produktkod. 30412464 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN67599510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Oxo-19-nor-17alpha-pregn-5(10)-en-20-yn-17-yl Acetate
CAS 19637-28-6
Molekylformel C22 H28 O3
Kvantitet 10 mg
Synonym 3-Oxo-19-nor-17alpha-pregn-5(10)-en-20-yn-17-yl Acetate, Norethisterone Acetate Imp. B (EP), (17α)-17-(Acetyloxy)-19-norpregn-5(10)-en-20-yn-3-one, 17-Hydroxy-19-Nor-17α-pregn-5(10)-en-20-yn-3-one Acetate, Norethindrone Acetate EP Impurity B
InChI formel InChI=1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,18-20H,5-13H2,2-3H3/t18-,19-,20+,21+,22+/m1/s1
LEDER CCOC([C@H](CCC1=CC=CC=C1)N(N=O)[C@@H](C)C(N2[C@@H](CCC2)C(O)=O)=O)=O
IUPAC-namn (8R,9S,13S,14S,17R)-17-Ethynyl-13-methyl-3-oxo-2,3,4,6,7,8,9,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl Acetate
Molekylvikt (g/mol) 340.46
Formel vikt 340.2