Learn More
5,12-Napthacenequinone, TRC
Product Code. 30471763
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30471763 1 g 1g
1 options
Product Code. 30471763 Supplier Toronto Research Chemicals Supplier No. TRCN3440001G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5,12-Napthacenequinone
CAS 1090-13-7
Molecular Formula C18H10O2
Quantity 1 g
Synonym Naphthacenequinone, 5,12-Naphthacenequinone, 5,12-Tetracenequinone, Benz[b]anthracene-5,12-dione, NSC 401183, Naphthacene-6,11-quinone, Tetracenemonoquinone
InChI Formula InChI=1S/C18H10O2/c19-17-13-7-3-4-8-14(13)18(20)16-10-12-6-2-1-5-11(12)9-15(16)17/h1-10H
SMILES O=C1c2ccccc2C(=O)c3cc4ccccc4cc13
IUPAC Name tetracene-5,12-dione
Molecular Weight (g/mol) 258.27
Formula Weight 258.0681