missing translation for 'onlineSavingsMsg'
Läs mer

5-(1H-Pyrrol-1-yl)-1H-benzo[d]imidazole-2-thiol, TRC

Produktkod. 30420262
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420262 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30420262 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP998833250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 5-(1H-Pyrrol-1-yl)-1H-benzo[d]imidazole-2-thiol
CAS 172152-53-3
Molekylformel C11H9N3S
Kvantitet 250 mg
Synonym Ilaprazole Impurity 61
InChI formel InChI=1S/C11H9N3S/c15-11-12-9-4-3-8(7-10(9)13-11)14-5-1-2-6-14/h1-7H,(H2,12,13,15)
LEDER Sc1nc2ccc(cc2[nH]1)n3cccc3
IUPAC-namn 6-pyrrol-1-yl-1H-benzimidazole-2-thiol
Molekylvikt (g/mol) 215.27
Formel vikt 215.0517
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.