Läs mer
5-((2-(2,2-Difluoroethoxy)-6-(trifluoromethyl)phenyl)sulfonamido)-4H-1,2,4-triazole-3-carboxylic Acid, TRC
Produktkod. 30368210
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30368210 1 mg
1 options
Produktkod. 30368210 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD3034951MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-((2-(2,2-Difluoroethoxy)-6-(trifluoromethyl)phenyl)sulfonamido)-4H-1,2,4-triazole-3-carboxylic Acid
CAS 907215-84-3
Molekylformel C12 H9 F5 N4 O5 S
Kvantitet 1 mg
Synonym 5-[[[2-(2,2-Difluoroethoxy)-6-(trifluoromethyl)phenyl]sulfonyl]amino]-1H-1,2,4-triazole-3-carboxylic acid, BSTCA, Penoxsulam-carboxylic Acid (BSTCA), Penoxsulam-carboxylic acid (BSTCA)
InChI formel InChI=1S/C12H9F5N4O5S/c13-7(14)4-26-6-3-1-2-5(12(15,16)17)8(6)27(24,25)21-11-18-9(10(22)23)19-20-11/h1-3,7H,4H2,(H,22,23)(H2,18,19,20,21)
LEDER OC(=O)c1nnc(NS(=O)(=O)c2c(OCC(F)F)cccc2C(F)(F)F)[nH]1
IUPAC-namn 5-[[2-(2,2-difluoroethoxy)-6-(trifluoromethyl)phenyl]sulfonylamino]-4H-1,2,4-triazole-3-carboxylic acid
Molekylvikt (g/mol) 416.28
Formel vikt 416.0214