Läs mer
5-[(2,5-Dioxo-1-pyrrolidinyl)methyl]-anastrozole, TRC
Produktkod. 30352460
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352460 500 mg
30365842 100 mg
30362391 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD241280500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1301724-97-9
Molekylformel C19H21N3O2
Molekylvikt (g/mol) 323.39
Synonym 5-[(2,5-Dioxo-1-pyrrolidinyl)methyl]-alpha1,alpha1,alpha3,alpha3-tetramethyl-1,3-benzenediacetonitrile, 2,2'-(5-((2,5-Dioxopyrrolidin-1-yl)methyl)-1,3-phenylene)bis(2-methylpropanenitrile)
LEDER CC(C)(C#N)c1cc(CN2C(=O)CCC2=O)cc(c1)C(C)(C)C#N
Specifikationer
Kemiskt namn eller material 5-[(2,5-Dioxo-1-pyrrolidinyl)methyl]-anastrozole
Kvantitet 500 mg
InChI formel InChI=1S/C19H21N3O2/c1-18(2,11-20)14-7-13(8-15(9-14)19(3,4)12-21)10-22-16(23)5-6-17(22)24/h7-9H,5-6,10H2,1-4H3
Formel vikt 323.1634