Läs mer
5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, TRC
Produktkod. 30358154
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358154 10 mg
30366558 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358154 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36965010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 898224-95-8
Molekylformel C18H26ClN3O3
Molekylvikt (g/mol) 367.87
Synonym Bendamustine Impurity
IUPAC-namn ethyl 4-(5-((2-chloroethyl)(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoate
LEDER OCCN(CCCl)C1=CC(N=C(CCCC(OCC)=O)N2C)=C2C=C1
Specifikationer
Kemiskt namn eller material 5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
Kvantitet 10 mg
InChI formel InChI=1S/C18H26ClN3O3/c1-3-25-18(24)6-4-5-17-20-15-13-14(7-8-16(15)21(17)2)22(10-9-19)11-12-23/h7-8,13,23H,3-6,9-12H2,1-2H3
Formel vikt 367.17