missing translation for 'onlineSavingsMsg'
Läs mer

5-[(2R)-2-Aminopropyl]-2-methoxybenzenesulfonamide, TRC

Produktkod. 30319433
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30319433 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30319433 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6286951G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 5-[(2R)-2-Aminopropyl]-2-methoxybenzenesulfonamide
CAS 112101-81-2
Molekylformel C10 H16 N2 O3 S
Kvantitet 1 g
Synonym Benzenesulfonamide, 5-[(2R)-2-aminopropyl]-2-methoxy-, Benzenesulfonamide, 5-(2-aminopropyl)-2-methoxy-, (R)-, 5-[(2R)-2-Aminopropyl]-2-methoxybenzenesulfonamide, (R)-(-)-5-(2-Aminopropyl)-2-methoxybenzenesulfonamide, (R)-5-(2-Aminopropyl)-2-methoxybenzenesulfonamide
InChI formel InChI=1S/C10H16N2O3S/c1-7(11)5-8-3-4-9(15-2)10(6-8)16(12,13)14/h3-4,6-7H,5,11H2,1-2H3,(H2,12,13,14)/t7-/m1/s1
LEDER COc1ccc(C[C@@H](C)N)cc1S(=O)(=O)N
IUPAC-namn 5-[(2R)-2-aminopropyl]-2-methoxybenzenesulfonamide
Molekylvikt (g/mol) 244.31
Formel vikt 244.0882
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.