Läs mer
5-(3-Chloropropyl)-1-phenyl-1H-1,2,3,4-tetrazole, TRC
Produktkod. 30366813
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366813 25 mg
30355799 250 mg
30350130 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366813 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC21467325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73963-34-5
Molekylformel C10H11N4Cl
Molekylvikt (g/mol) 222.67
IUPAC-namn 5-(3-chloropropyl)-1-phenyltetrazole
LEDER ClCCCc1nnnn1c2ccccc2
Specifikationer
Kemiskt namn eller material 5-(3-Chloropropyl)-1-phenyl-1H-1,2,3,4-tetrazole
Kvantitet 25 mg
InChI formel InChI=1S/C10H11ClN4/c11-8-4-7-10-12-13-14-15(10)9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2
Formel vikt 222.0672