Läs mer
5-(3-Chloropropyl)-5H-dibenz[b,f]azepine, Mikromol™
Produktkod. 30049282
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049282 25 mg
30051846 100 mg
30462353 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049282 Leverantör Mikromol Leverantörsnummer MM0422.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(3-Chloropropyl)-5H-dibenz[b,f]azepine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-(3-Chloropropyl)-5H-dibenz[b,f]azepine
CAS 51551-40-7
Analyt- eller komponentnamn 5-(3-Chloropropyl)-5H-dibenz[b,f]azepine
Frakt skick Room Temperature
Molekylformel C17 H16 Cl N
Certifikat och efterlevnad ISO 9001
Synonym 5-(3-Chloropropyl)-5H-dibenz[b,f]azepine
InChI formel InChI=1S/C17H16ClN/c18-12-5-13-19-16-8-3-1-6-14(16)10-11-15-7-2-4-9-17(15)19/h1-4,6-11H,5,12-13H2
LEDER ClCCCN1c2ccccc2C=Cc3ccccc13
IUPAC-namn 11-(3-chloropropyl)benzo[b][1]benzazepine
Molekylvikt (g/mol) 269.77
Kvantitet 25 mg
Formel vikt 269.0971 g/mol