missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-(3-Phenoxyphenyl)-1H-tetrazole, 98%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 152°C to 154°C |
Quantity | 250 mg |
Formula Weight | 238.25 |
Percent Purity | 98% |
Chemical Name or Material | 5-(3-Phenoxyphenyl)-1H-tetrazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'