Läs mer
5-[[4-[2-(5-Ethyl-2-pyridinyl)ethoxy]phenyl]-methylene]-2,4-thiazolidinedione(Pioglitazone Impurity), TRC
Produktkod. 30136139
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136139 50 mg
30136283 25 mg
30010971 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92559050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144809-28-9
Molekylformel C19 H18 N2 O3 S
Molekylvikt (g/mol) 354.42
Synonym 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)ethoxy]phenyl]methylene]-, 5-[[4-[2-(5-Ethyl-2-pyridyl)ethoxy]phenyl]methylene]thiazolidine-2,4-dione
IUPAC-namn (5E)-5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methylidene]-1,3-thiazolidine-2,4-dione
LEDER CCc1ccc(CCOc2ccc(\C=C/3\SC(=O)NC3=O)cc2)nc1
Specifikationer
Kemiskt namn eller material 5-[(4-[2-(5-Ethyl-2-pyridinyl)ethoxy]phenyl)methylene]-2,4-thiazolidinedione
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C19H18N2O3S/c1-2-13-3-6-15(20-12-13)9-10-24-16-7-4-14(5-8-16)11-17-18(22)21-19(23)25-17/h3-8,11-12H,2,9-10H2,1H3,(H,21,22,23)/b17-11+
Formel vikt 354.1038
Procent renhet >95%