missing translation for 'onlineSavingsMsg'
Learn More

5-[4-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole, 95%, Thermo Scientific™

Catalog Number p-3522546
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
362529-03-1
C10H9BrN2O
253.099
MFCD09025866
RWTXHMRWDVQEOV-UHFFFAOYSA-N
5-4-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-4-bromomethyl phenyl-3-methyl, 1,2,4-oxadiazole,5-4-bromomethyl phenyl-3-methyl
18374480
5-[4-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
CC1=NOC(=N1)C2=CC=C(C=C2)CBr
This item is not returnable. View return policy
362529-03-1
C10H9BrN2O
253.099
MFCD09025866
RWTXHMRWDVQEOV-UHFFFAOYSA-N
5-4-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-4-bromomethyl phenyl-3-methyl, 1,2,4-oxadiazole,5-4-bromomethyl phenyl-3-methyl
18374480
5-[4-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
CC1=NOC(=N1)C2=CC=C(C=C2)CBr
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 253.1
Percent Purity 95%
Chemical Name or Material 5-[4-(Bromomethyl)phenyl]-3-methyl-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'