missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic Acid, TRC

Product Code. 30349267
Change view
Click to view available options
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30349267 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349267 Supplier Toronto Research Chemicals Supplier No. TRCC37783010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic Acid
CAS 162758-35-2
Molekylformel C17H11Cl3N2O2
Kvantitet 10 g
Synonym 1-(2,4-Dichlorophenyl)-5-(4-chlorophenyl)-4-methyl-1H-pyrazole-3-carboxylic Acid, 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxylic Acid
InChI formel InChI=1S/C17H11Cl3N2O2/c1-9-15(17(23)24)21-22(14-7-6-12(19)8-13(14)20)16(9)10-2-4-11(18)5-3-10/h2-8H,1H3,(H,23,24)
LEDER Cc1c(nn(c2ccc(Cl)cc2Cl)c1c3ccc(Cl)cc3)C(=O)O
IUPAC-namn 5-(4-chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid
Molekylvikt (g/mol) 381.64
Formel vikt 379.9886
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.