Läs mer
5-(4-Nitrophenyl)furfural, TRC
Produktkod. 30479424
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479424 5 g
30474421 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479424 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5035355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7147-77-5
Molekylformel C11 H7 N O4
Molekylvikt (g/mol) 217.18
Synonym 5-(4-Nitrophenyl)furan-2-carbaldehyde, 5-(4-Nitrophenyl)-2-furancarboxyaldehyde
IUPAC-namn 5-(4-nitrophenyl)furan-2-carbaldehyde
LEDER [O-][N+](=O)c1ccc(cc1)c2oc(C=O)cc2
Specifikationer
Kemiskt namn eller material 5-(4-nitrophenyl)-2-furancarboxyaldehyde
Kvantitet 5 g
InChI formel InChI=1S/C11H7NO4/c13-7-10-5-6-11(16-10)8-1-3-9(4-2-8)12(14)15/h1-7H
Formel vikt 217.0375