missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Bromo-2-thienyl)isoxazole, ≥97%, Thermo Scientific™

Catalog Number p-3110770
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
138716-31-1
C7H4BrNOS
230.079
MFCD03618406
FCUZGSQWFAXBSB-UHFFFAOYSA-N
2-bromo-5-isoxazol-5-yl thiophene, 5-5-bromo-2-thienyl isoxazole, 5-5-bromothiophen-2-yl-1,2-oxazole, 5-bromo-2-isoxazol-5-ylthiophene, 5-5-bromothien-2-yl isoxazole, 5-5-bromo-2-thiophenyl isoxazole, 5-5-bromothiophen-2-yl isoxazole, 5-5-bromothien-2-yl-1,2-oxazole, 5-5-bromanylthiophen-2-yl-1,2-oxazole
2757026
5-(5-bromothiophen-2-yl)-1,2-oxazole
C1=C(SC(=C1)Br)C2=CC=NO2
This item is not returnable. View return policy
138716-31-1
C7H4BrNOS
230.079
MFCD03618406
FCUZGSQWFAXBSB-UHFFFAOYSA-N
2-bromo-5-isoxazol-5-yl thiophene, 5-5-bromo-2-thienyl isoxazole, 5-5-bromothiophen-2-yl-1,2-oxazole, 5-bromo-2-isoxazol-5-ylthiophene, 5-5-bromothien-2-yl isoxazole, 5-5-bromo-2-thiophenyl isoxazole, 5-5-bromothiophen-2-yl isoxazole, 5-5-bromothien-2-yl-1,2-oxazole, 5-5-bromanylthiophen-2-yl-1,2-oxazole
2757026
5-(5-bromothiophen-2-yl)-1,2-oxazole
C1=C(SC(=C1)Br)C2=CC=NO2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 230.08
Percent Purity 97%
Chemical Name or Material 5-(5-Bromo-2-thienyl)isoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'