missing translation for 'onlineSavingsMsg'
Läs mer

5,5-Diethoxypentanal, TRC

Produktkod. 30374584
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374584 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30374584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44192510MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 5,5-Diethoxypentanal
CAS 79893-96-2
Molekylformel C9 H18 O3
Kvantitet 10 mg
InChI formel InChI=1S/C9H18O3/c1-3-11-9(12-4-2)7-5-6-8-10/h8-9H,3-7H2,1-2H3
LEDER CCOC(CCCC=O)OCC
IUPAC-namn 5,5-diethoxypentanal
Molekylvikt (g/mol) 174.24
Formel vikt 174.1256
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.