Läs mer
5,6,7,8-tetrahydroquinazoline-6-carboxylic acid, TRC
Produktkod. 30454253
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454253 10 mg
30454225 100 mg
30452608 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454253 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30420510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1369096-46-7
Molekylformel C9H10N2O2
Molekylvikt (g/mol) 178.19
IUPAC-namn 5,6,7,8-tetrahydroquinazoline-6-carboxylic acid
LEDER OC(=O)C1CCc2ncncc2C1
Specifikationer
Kemiskt namn eller material 5,6,7,8-tetrahydroquinazoline-6-carboxylic acid
Kvantitet 10 mg
InChI formel InChI=1S/C9H10N2O2/c12-9(13)6-1-2-8-7(3-6)4-10-5-11-8/h4-6H,1-3H2,(H,12,13)
Formel vikt 178.0742