Läs mer
5(6)-Carboxytetramethylrhodamine Succinimidyl Ester (Technical Grade), TRC
Produktkod. 30359092
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359092 25 mg
30358460 10 mg
30367042 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18265025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 246256-50-8
Molekylformel C24 H22 N2 O3 . C6 H7 N O4
Molekylvikt (g/mol) 543.567
Synonym 5,6-Carboxytetramethylrhodamine Succinimidyl Ester, 5/6-TAMRA-SE, NHS-Rhodamine
IUPAC-namn 2-[3,6-bis(dimethylamino)xanthen-10-ium-9-yl]benzoate;(2,5-dioxopyrrolidin-1-yl) acetate
LEDER CN(C)c1ccc2c(c3ccccc3C(=O)[O-])c4ccc(cc4[o+]c2c1)N(C)C.CC(=O)ON5C(=O)CCC5=O
Specifikationer
Kemiskt namn eller material 5(6)-Carboxytetramethylrhodamine Succinimidyl Ester (Technical Grade)
Kvantitet 25 mg
InChI formel InChI=1S/C24H22N2O3.C6H7NO4/c1-25(2)15-9-11-19-21(13-15)29-22-14-16(26(3)4)10-12-20(22)23(19)17-7-5-6-8-18(17)24(27)28;1-4(8)11-7-5(9)2-3-6(7)10/h5-14H,1-4H3;2-3H2,1H3
Formel vikt 543.201