Läs mer
5,6-Dehydro-17Beta-dutasteride, TRC
Produktkod. 30353032
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353032 5 mg
30358002 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353032 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2295605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1430804-85-5
Molekylformel C27 H28 F6 N2 O2
Molekylvikt (g/mol) 526.51
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10-dodecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS)-, Dutasteride Imp. G (EP), Dutasteride 5-ene (USP), 5,6-Dehydro-17β-dutasteride
IUPAC-namn (1S,3aS,3bS,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,6,9b,10,11-decahydroindeno[5,4-f]quinoline-1-carboxamide
LEDER C[C@]12CC[C@H]3[C@@H](CC=C4NC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)Nc5cc(ccc5C(F)(F)F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 5,6-dehydro-17beta-dutasteride
Kvantitet 5 mg
InChI formel InChI=1S/C27H28F6N2O2/c1-24-11-9-17-15(4-8-21-25(17,2)12-10-22(36)35-21)16(24)6-7-19(24)23(37)34-20-13-14(26(28,29)30)3-5-18(20)27(31,32)33/h3,5,8,10,12-13,15-17,19H,4,6-7,9,11H2,1-2H3,(H,34,37)(H,35,36)/t15-,16-,17-,19+,24-,25+/m0/s1
Formel vikt 526.2055