Läs mer
5,6-Dehydro Finasteride, TRC
Produktkod. 30012464
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012464 50 mg
30010628 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012464 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22965550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1800205-94-0
Molekylformel C23 H34 N2 O2
Molekylvikt (g/mol) 370.53
Synonym N-(1,1-Dimethylethyl)-3-oxo-4-azaandrosta-1,5-diene-17β-carboxamide, (4aR,4bS,6aS,7S,9aS,9bS)-N-(1,1-Dimethylethyl)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10-dodecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, 5,6-Dehydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10-dodecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS)-, N-(1,1-Dimethylethyl)-3-oxo-4-azaandrosta-1,5-diene-17beta-carboxamide, (4aR,4bS,6aS,7S,9aS,9bS)-N-(tert-butyl)-4a,6a-dimethyl-2-oxo-2,4a,4b,5,6,6a,7,8,9,9a,9b,10-dodecahydro-1H-indeno[5,4-f]quinoline-7-carboxamide
IUPAC-namn (1S,3aS,3bS,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,6,9b,10,11-decahydroindeno[5,4-f]quinoline-1-carboxamide
LEDER CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Finasteride EP Impurity C
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C23H34N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h9,11,13-17H,6-8,10,12H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,22-,23+/m0/s1
Formel vikt 370.262
Procent renhet >95%