Läs mer
5,6-Diamino-2,4-dihydroxypyrimidine, Hydrochloride Salt, TRC
Produktkod. 30355946
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30355946 1 g
1 options
Produktkod. 30355946 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4163351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5,6-Diamino-2,4-dihydroxypyrimidine, Hydrochloride Salt
CAS 53608-89-2
Molekylformel C4 H6 N4 O2 . H Cl
Kvantitet 1 g
Synonym 5,6-Diaminouracil Hydrochloride, 2,4(1H,3H)-Pyrimidinedione, 5,6-diamino-, hydrochloride (1:1), 2,4(1H,3H)-Pyrimidinedione, 5,6-diamino-, monohydrochloride (9CI), 4,5-Diamino-2,6-dihydroxypyrimidine hydrochloride, 5,6-Diaminouracil hydrochloride
InChI formel InChI=1S/C4H6N4O2.ClH/c5-1-2(6)7-4(10)8-3(1)9;/h5H2,(H4,6,7,8,9,10);1H
LEDER Cl.NC1=C(N)C(=O)NC(=O)N1
IUPAC-namn 5,6-diamino-1H-pyrimidine-2,4-dione;hydrochloride
Molekylvikt (g/mol) 178.577
Formel vikt 178.0258