Läs mer
5,6-Diamino-2-hydroxybenzimidazole Dihydrochloride, TRC
Produktkod. 30356253
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30356253 25 mg
1 options
Produktkod. 30356253 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41634025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5,6-Diamino-2-hydroxybenzimidazole, Dihydrochloride
CAS 42815-81-6
Molekylformel C7 H8 N4 O . 2 Cl H
Kvantitet 25 mg
Synonym 2H-Benzimidazol-2-one, 5,6-diamino-1,3-dihydro-, hydrochloride (1:2), 2H-Benzimidazol-2-one, 5,6-diamino-1,3-dihydro-, dihydrochloride (9CI), 5,6-Diamino-2(3H)-benzimidazolone dihydrochloride, 5,6-Diaminobenzimidazolone dihydrochloride, 5,6-Diamino-2-hydroxybenzimidazole dihydrochloride
InChI formel InChI=1S/C7H8N4O.2ClH/c8-3-1-5-6(2-4(3)9)11-7(12)10-5;;/h1-2H,8-9H2,(H2,10,11,12);2*1H
LEDER Cl.Cl.Nc1cc2nc(O)[nH]c2cc1N
IUPAC-namn 5,6-diamino-1H-benzimidazol-2-ol;dihydrochloride
Molekylvikt (g/mol) 237.09
Formel vikt 236.0232