missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diaminopyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11906471
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 1204334-21-3
Melting Point 198°C to 202°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C11H18BN3O2
MDL Number MFCD11054043
Quantity 1 g
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2,3-diamine, 2,3-diaminopyridine-5-boronic acid, pinacol ester, 5,6-diaminopyridine-3-boronic acid pinacol ester, 2,3-diaminopyridine-5-boronic acid pinacol ester, 5,6-diaminopyridin-3-ylboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2,3-diamine
InChI Key XYAJKHYERTYNDS-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC(N)=C(N)N=C1
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2,3-diamine
Molecular Weight (g/mol) 235.09
PubChem CID 46738008
Formula Weight 235.1
Percent Purity 97%
Chemical Name or Material 5, 6-Diaminopyridine-3-boronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 5,6-Diaminopyridine-3-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'