Läs mer
5,6-Dichloronicotinic acid, 98+%, Thermo Scientific Chemicals
Produktkod. 11427266
Change view
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11427266 5 g 5g
11437266 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11427266 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B25586.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41667-95-2
Molekylformel C6H3Cl2NO2
Molekylvikt (g/mol) 191.995
MDL-nummer MFCD00075181
InChI-nyckel RNRLTTNKVLFZJS-UHFFFAOYSA-N
Synonym 5,6-dichloronicotinic acid, 5,6-dichloro nicotinic acid, 5,6-dichloro-3-pyridinecarboxylic acid, 5,6-dichloronicotinicacid, 2,3-dichloro-5-carboxypyridine, 3-pyridinecarboxylic acid, 5,6-dichloro, 5,6-dichloro-nicotinic acid, pubchem12968, acmc-209jl4, 5,6 dichloronicotinic acid
PubChem CID 247987
IUPAC-namn 5,6-dichloropyridine-3-carboxylic acid
LEDER C1=C(C=NC(=C1Cl)Cl)C(=O)O
Specifikationer
Kemiskt namn eller material 5,6-Dichloronicotinic acid
Smältpunkt ∼164°C (decomposition)
Kvantitet 5 g
Beilstein 383740
Formel vikt 192
Procent renhet ≥98%