Läs mer
5,6-Dichloronicotinic acid, TRC
Produktkod. 30376825
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30376825 500 mg
1 options
Produktkod. 30376825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD435653500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5,6-Dichloronicotinic acid
CAS 41667-95-2
Molekylformel C6H3Cl2NO2
Kvantitet 500 mg
Synonym 5,6-Dichloro-3-pyridinecarboxylic Acid, 2,3-Dichloropyridine-5-carboxylic Acid, 5,6-Dichloronicotinic Acid, 5,6-Dichloropyridine-3-carboxylic Acid, NSC 63885
InChI formel InChI=1S/C6H3Cl2NO2/c7-4-1-3(6(10)11)2-9-5(4)8/h1-2H,(H,10,11)
LEDER NC1=CC=CC=C1C(C2=CC=CC=C2Br)=O
Molekylvikt (g/mol) 192