Läs mer
5,6-Dihydro-3-(methylamino)-2H-1,4-thiazin-2-one Oxime Hydrochloride, TRC
Produktkod. 30370014
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30370014 100 mg
1 options
Produktkod. 30370014 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD449435100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(Methylamino)-5,6-dihydro-2H-1,4-thiazin-2-one Oxime Hydrochloride
CAS 112233-23-5
Molekylformel C5 H9 N3 O S . Cl H
Kvantitet 100 mg
Synonym 2H-1,4-Thiazin-2-one, 5,6-dihydro-3-(methylamino)-, oxime, hydrochloride (1:1), 2H-1,4-Thiazin-2-one, 5,6-dihydro-3-(methylamino)-, oxime, monohydrochloride (9CI), 3-(Methylamino)-5,6-dihydro-2H-1,4-thiazin-2-one Oxime Hydrochloride, AH 22888A, Ph Eur Ranitidine Impurity G, Ranitidine oxime, hydrochloride salt (AH22888), oxime (AH 22888A)
InChI formel InChI=1S/C5H9N3OS.ClH/c1-6-4-5(8-9)10-3-2-7-4;/h9H,2-3H2,1H3,(H,6,7);1H
LEDER Cl.CNC1=NCCSC1=NO
IUPAC-namn N-[5-(methylamino)-2,3-dihydro-1,4-thiazin-6-ylidene]hydroxylamine;hydrochloride
Molekylvikt (g/mol) 195.67
Formel vikt 195.023