Läs mer
5,6-Dihydro Uracil-13C,15N2, TRC
Produktkod. 30008208
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30008208 2.5 mg 2.5mg
30010548 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008208 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4499922.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 181516-96-1
Molekylformel C313CH615N2O2
Molekylvikt (g/mol) 117.08
Synonym Dihydro-2,4(1H,3H)-Pyrimidinedione-2-13C-1,3-15N2
LEDER O=C(CC[15N]1[H])[15NH][13C]1=O
Specifikationer
Kemiskt namn eller material 5,6-Dihydro Uracil-13C,15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C4H6N2O2/c7-3-1-2-5-4(8)6-3/h1-2H2,(H2,5,6,7,8)/i4+1,5+1,6+1
Formel vikt 117.0404
Procent renhet >95%